Skip to Content
Merck
All Photos(1)

Key Documents

W287105

Sigma-Aldrich

Phenethyl isovalerate

≥98%, FCC, FG

Synonym(s):

Benzyl carbinyl isovalerianate, Phenethyl isopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO2CH2CH2C6H5
CAS Number:
Molecular Weight:
206.28
FEMA Number:
2871
EC Number:
Council of Europe no.:
461
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.466
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.485 (lit.)

bp

268 °C (lit.)

density

0.974 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; rose; sweet

SMILES string

CC(C)CC(=O)OCCc1ccccc1

InChI

1S/C13H18O2/c1-11(2)10-13(14)15-9-8-12-6-4-3-5-7-12/h3-7,11H,8-10H2,1-2H3

InChI key

JIMGVOCOYZFDKB-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justyna Aszyk et al.
Journal of chromatography. A, 1547, 86-98 (2018-03-15)
Many flavour compounds that are present in e-liquids for e-cigarettes are responsible for specific tastes and smoking sensations for users. Data concerning content and specific types of flavours is often limited and unknown to users. The aim of the research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service