Skip to Content
Merck
All Photos(1)

Key Documents

W268208

Sigma-Aldrich

Methyl anthranilate

≥98%, FCC, FG

Synonym(s):

Methyl 2-aminobenzoate, Methyl anthranilate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(H2N)C6H4CO2CH3
CAS Number:
Molecular Weight:
151.16
FEMA Number:
2682
Beilstein:
606965
EC Number:
Council of Europe no.:
250
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.715
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor pressure

1 mmHg ( 20 °C)

Assay

≥98%

autoignition temp.

986 °F

refractive index

n20/D 1.582 (lit.)

bp

256 °C (lit.)

mp

24 °C (lit.)

density

1.168 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

grape; musty; fruity; sweet; wine-like

SMILES string

COC(=O)c1ccccc1N

InChI

1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3

InChI key

VAMXMNNIEUEQDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl anthranilate occurs naturally in grapes[1] and strawberries.[2] It is also found in Burgundy Pinot noir wines[3], citrus honey[4] and American wines.[5] Methyl anthranilate is also a potent bird repellant.[6]

Application

Methyl anthranilate is a sunscreen precursor (an ortho compound) used as an odor compound in the study of controlled odor release from the cavity of the cyclodextrin.[7][8]

Biochem/physiol Actions

Taste at 2-5 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Volatiles from grapes I. Some volatiles from concord essence.
Stevens KL, et al.
Journal of Food Science, 30(6), 1006-1007 (1965)
The volatiles of two new strawberry cultivars,?Annelie? and ?Alaska Pioneer?, obtained by backcrossing of cultivated strawberries with wild strawberries, fragaria vesca, rugen and fragaria virginiana.
Hirvi T & Honkanen E.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 175(2), 113-116 (1982)
Methyl anthranilate as an aroma constituent of American wine.
Nelson RR, et al.
Journal of Food Science, 42(1), 57-59 (1977)
Visualization of controlled fragrance release from cyclodextrin inclusion complexes by fluorescence imaging
Liu C and Hayashi K
Flavour and Fragrance Journal, 29(6), 356-363 (2014)
A comparative study of hesperetin and methyl anthranilate as markers of the floral origin of citrus honey.
Ferreres F, et al.
Journal of the Science of Food and Agriculture, 65(3), 371-372 (1994)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service