Skip to Content
Merck
All Photos(1)

Key Documents

W267007

Sigma-Aldrich

p-Anisaldehyde

≥97.5%, FCC, FG

Synonym(s):

4-Methoxybenzaldehyde, Aubépine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2670
Beilstein:
471382
EC Number:
Council of Europe no.:
103
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.015
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.515

vapor density

4.7 (vs air)

Assay

≥97.5%

form

liquid

composition

contains IFRA restricted p-Anisaldehyde

refractive index

n20/D 1.573 (lit.)

bp

248 °C (lit.)

mp

−1 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

anise; cherry; creamy; floral; balsamic; sweet; vanilla

SMILES string

[H]C(=O)c1ccc(OC)cc1

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application


  • Aryl-alcohol oxidase involved in lignin degradation: a mechanistic study based on steady and pre-steady state kinetics and primary and solvent isotope effects with two alcohol substrates.: The study focuses on the mechanistic understanding of aryl-alcohol oxidase in lignin degradation, utilizing p-Anisaldehyde as a model substrate to investigate enzyme kinetics and isotope effects. (Ferreira et al., 2009).

Biochem/physiol Actions

Odor at 1.0%
Taste at 5-10 ppm

Other Notes

Natural occurrence: Vanilla, fennel, star anise, cranberry, black currant, cinnamon, basil.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Juillet et al.
Plant biology (Stuttgart, Germany), 13(4), 570-575 (2011-06-15)
It has often been proposed that nectarless deceptive orchid species exploit naïve pollinators in search of food before they learn to avoid their flowers, and that intraspecific floral trait polymorphism, often noted in this plant group, could prolong the time
Israt Jahan et al.
Acta pharmaceutica (Zagreb, Croatia), 60(2), 207-215 (2010-12-08)
In this study, five compounds, lupeol (1), epilupeol (2), β-sitosterol (3), stigmasterol (4) and p-methoxybenzaldehyde (5) were isolated from the petroleum ether and dichloromethane fractions of a methanolic extract of the stem bark of Delonix regia. Antimicrobial screening of the
Guozhu Zhang et al.
Journal of the American Chemical Society, 130(38), 12598-12599 (2008-08-30)
A novel approach to generate Au-containing all-carbon 1,3-dipoles is developed via an unprecedented migration-fragmentation of ketals/acetals. These in situ generated dipoles undergo rapid [3+2] cycloaddition under mild conditions with various enones/enals, electron-rich aromatic aldehydes, and N-benzylindole, leading to flexible formation
Tae Joung Ha et al.
Journal of agricultural and food chemistry, 53(18), 7024-7028 (2005-09-01)
Anisaldehyde (p-methoxybenzaldehyde) was previously reported to inhibit the tyrosinase-catalyzed oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) noncompetitively as long as the enzyme activity was monitored by measuring dopachrome formation. However, anisaldehyde did not inhibit this oxidation if a longer reaction time was observed
Wenxin Gu et al.
Organic letters, 5(4), 415-418 (2003-02-14)
[reaction: see text] Two new 4-methoxybenzaldehyde backbone linker resins were developed for the solid-phase synthesis of peptides. The linkers are very stable during the cleavage of common protecting groups for amines (Fmoc, Boc) and carboxylic acids (Me, All, tBu) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service