Skip to Content
Merck
All Photos(3)

Key Documents

W250716

Sigma-Aldrich

Geraniol

greener alternative
Geraniol

natural, ≥97%, FG

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2OH
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2507
Beilstein:
1722456
Council of Europe no.:
60
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.012
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.60

vapor density

5.31 (vs air)

Assay

≥97%

composition

contains IFRA and EU 1223/2009 restricted Geraniol

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

impurities

<30% β-citronellol

refractive index

n20/D 1.474 (lit.)

bp

229-230 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

geraniol

greener alternative category

Organoleptic

citrus; floral; fruity; rose; sweet

SMILES string

C\C(C)=C\CC\C(C)=C\CO

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

GLZPCOQZEFWAFX-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Geraniol is a monoterpene alcohol that occurs as one of the main flavor constituents in Kansho-shochu[1], sweet basil[2], hopped beer[3] and green tea.[4]
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Pectin/sodium alginate-based active film integrated with microcrystalline cellulose and geraniol for food packaging applications.: Investigates the use of geraniol in developing biodegradable active packaging films, emphasizing its role in enhancing antimicrobial properties (Hoque et al., 2024).

  • The enhancement of flowery-like aroma in green tea under optimized processing conditions by sensory-directed flavor analysis.: Explores how geraniol enhances the flowery-like aroma in green tea, with a focus on sensory evaluation and optimization of processing conditions (Wang et al., 2024).

  • Antifungal activity of bio-active cell-free culture extracts and volatile organic compounds (VOCs) synthesised by endophytic fungal isolates of Garden Nasturtium.: Discusses the synthesis of VOCs including geraniol by fungal isolates, highlighting their antifungal activities against plant pathogens (Santra et al., 2024).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristic flavor of Kamho-shochu (sweet potato spirit).
Ohta T, et al.
Agricultural and Biological Chemistry, 54(6), 1353-1357 (1990)
Biotransformation of hop-derived monoterpene alcohols by lager yeast and their contribution to the flavor of hopped beer.
Takoi K, et al.
Journal of Agricultural and Food Chemistry, 58(8), 5050-5058 (2010)
Characterization of geraniol synthase from the peltate glands of sweet basil.
Iijima Y, et al.
Plant Physiology, 134(1), 370-379 (2004)
Antimicrobial activity of green tea flavor components and their combination effects.
Kubo I, et al.
Journal of Agricultural and Food Chemistry, 40(2), 245-248 (1992)
Gavin L Sacks et al.
Journal of agricultural and food chemistry, 60(12), 2998-3004 (2012-03-09)
1,1,6-Trimethyl-1,2-dihydronaphthalene (TDN) is well-known to contribute "petrol" aromas to aged Riesling wines, but its prevalence and contribution to young Riesling or non-Riesling wines is not well understood. TDN concentrations were measured in 1-3-year-old varietal wines produced from Cabernet franc (n

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service