Skip to Content
Merck
All Photos(1)

Key Documents

W222003

Sigma-Aldrich

Isobutyraldehyde

≥98%, FG

Synonym(s):

2-Methylpropanal, Isobutanal, 2-Methylpropionaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11
FEMA Number:
2220
Beilstein:
605330
EC Number:
Council of Europe no.:
92
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.004
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

2.5 (vs air)

vapor pressure

66 mmHg ( 4.4 °C)

Assay

≥98%

autoignition temp.

384 °F

expl. lim.

10 %, 25 °F
2 %, 32 °F

refractive index

n20/D 1.374 (lit.)

bp

63 °C (lit.)

mp

−65 °C (lit.)

solubility

60 g/L at 25 °C

density

0.79 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; floral

storage temp.

2-8°C

SMILES string

[H]C(=O)C(C)C

InChI

1S/C4H8O/c1-4(2)3-5/h3-4H,1-2H3

InChI key

AMIMRNSIRUDHCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyraldehyde is an aliphatic aldehyde that can be used as a flavoring agent.[1] It has been identified as one of the volatile flavor components in wheat essence.[2] Some humans are anosmic towards the odor of isobutanal.[3]

Application


  • Single-Atom palladium engineered cobalt nanocomposite for selective aerobic oxidation of sulfides to sulfoxides.: This study presents a novel approach using a palladium-engineered cobalt nanocomposite that promotes the selective aerobic oxidation of sulfides, with isobutyraldehyde acting as an important reagent in the process, offering potential for more efficient and environmentally friendly chemical synthesis (Ma et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-11.2 °F - closed cup

Flash Point(C)

-24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 1462-1463 (1997)
Wheat flavor components
Mcwilliams M & Mackey AC
Journal of Food Science, 34(6), 493-496 (1969)
Specific anosmia to isobutyraldehyde: the malty primary odor.
MOORE JE, et al.
Chemical Senses, 2(1), 17-25 (1976)
Rudy Coquet et al.
Physical chemistry chemical physics : PCCP, 9(45), 6040-6046 (2007-11-16)
The formation and catalytic behavior of active structures in a SiO(2)-supported unsaturated Ru complex catalyst for alkene epoxidation were studied by means of hybrid density functional theory (DFT) calculations. An energy-gaining route for the catalyst activation was found to allow
Christine Counet et al.
Journal of agricultural and food chemistry, 50(8), 2385-2391 (2002-04-04)
After vacuum distillation and liquid-liquid extraction, the volatile fractions of dark chocolates were analyzed by gas chromatography-olfactometry and gas chromatography-mass spectrometry. Aroma extract dilution analysis revealed the presence of 33 potent odorants in the neutral/basic fraction. Three of these had

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service