Skip to Content
Merck
All Photos(3)

Key Documents

W215311

Sigma-Aldrich

Bergamot oil

greener alternative

colorless, natural, product of Simone Gatto

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2153
MDL number:
UNSPSC Code:
12164502

biological source

citrus (Bergamia Risso et Poiteau (Fam. Rutaceae))

Quality Level

grade

Fragrance grade
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

composition

contains IFRA and EU 1223/2009 restricted Citral, Citronellol, Limonene, Linalool and Citrus bergamia

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

color

colorless

refractive index

n20/D 1.4665 (lit.)
n20/D 1.4675 (lit.)

bp

159 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

limonene (sum of D, L and DL), linalool, citral (neral+geranial), citronellol, Citrus bergamia peel oil expressed

greener alternative category

Organoleptic

orange; citrus; woody

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service