TEM-LFH7100AR1.3
QUANTIFOIL on London Finder Grids (H7)
Gold Grids, size 400 mesh, pack of 100
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Recommended Products
packaging
pack of 100
size
400 mesh
Related Categories
General description
QUANTIFOIL Gold Grids, 200 Mesh, 1.2 micron hole, 2.5 micron period. London Finder Grids (H7), Pitch 63 micron, 400 mesh
Choose from one of the most recent versions:
Certificates of Analysis (COA)