Skip to Content
Merck
All Photos(1)

Key Documents

T511293

Sigma-Aldrich

(2′S)-2′-Deoxy-2′-fluoro-5-ethynyluridine, (F-ara-EdU)

AldrichCPR

Synonym(s):

1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-ethynyl-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11FN2O5
CAS Number:
Molecular Weight:
270.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

reaction suitability

reaction type: click chemistry

SMILES string

OC[C@H]1O[C@H]([C@@H](F)[C@@H]1O)N2C=C(C#C)C(=O)NC2=O

InChI

1S/C11H11FN2O5/c1-2-5-3-14(11(18)13-9(5)17)10-7(12)8(16)6(4-15)19-10/h1,3,6-8,10,15-16H,4H2,(H,13,17,18)/t6-,7+,8-,10-/m1/s1

InChI key

YEEGMPUOCRQFRV-IBCQBUCCSA-N

Application

A probe for DNA synthesis in vivo that is less toxic than both EdU and BrdU. Ideally suited for metabolic DNA labeling when prolonged cell survival and undisturbed cell cycle progression are required.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne B Neef et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(51), 20404-20409 (2011-12-07)
Commonly used metabolic labels for DNA, including 5-ethynyl-2'-deoxyuridine (EdU) and BrdU, are toxic antimetabolites that cause DNA instability, necrosis, and cell-cycle arrest. In addition to perturbing biological function, these properties can prevent metabolic labeling studies where subsequent tissue survival is
Florence D D'Orazi et al.
The Journal of comparative neurology, 528(17), 2816-2830 (2020-04-29)
A major challenge in regenerative medicine is replacing cells lost through injury or disease. While significant progress has been made, much remains unknown about the accuracy of native regenerative programs in cell replacement. Here, we capitalized on the regenerative capacity
Lingluo Chu et al.
Molecular cell, 79(6), 902-916 (2020-08-10)
A long-standing conundrum is how mitotic chromosomes can compact, as required for clean separation to daughter cells, while maintaining close parallel alignment of sister chromatids. Pursuit of this question, by high resolution 3D fluorescence imaging of living and fixed mammalian
Sameer S Chopra et al.
Cell systems, 10(1), 66-81 (2019-12-10)
Frequent mutation of PI3K/AKT/mTOR signaling pathway genes in human cancers has stimulated large investments in targeted drugs but clinical successes are rare. As a result, many cancers with high PI3K pathway activity, such as triple-negative breast cancer (TNBC), are treated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service