Documents
SYNV210250
Synthware Dewar type vacuum trap
250 mL, stopcock: PTFE
Synonym(s):
Synthware V210250
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V210250
capacity
250 mL
General description
Dry ice, Dewar style trap condenser designed for large volume solvent removal. Choice either glass stopcock for better vacuum or PTFE stopcock for grease-free operation. With 10 mm hose connections.
Legal Information
Synthware is a trademark of Kemtech America Inc.