SYNS351656
Synthware™ high vacuum double oblique glass stopcock with hollow plug
bore 4 mm, stem O.D. 10 mm, Plug Size: 16.2/56
Synonym(s):
Synthware S351656
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
S351656
bore
4 mm
stem O.D.
10 mm
Related Categories
General description
Synthware™ double oblique bore glass stopcock has diagonal, parallel bores that allow the connection of a single stem to both stems on the opposite side. The hollow glass plug option is precision ground with corresponding numbers on the barrel and plug. The lower vacuum chamber is evacuated via a separate 2 mm hole in the plug, which is not in the line of flow.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)