Key Documents
SYNF442250
Synthware three neck round bottom flask with vertical side necks
250 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22
Synonym(s):
Synthware F442250, 3-neck round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Pricing and availability is not currently available.
material
borosilicate glass flask
round bottom
three-neck (vertical side necks)
Joint
center joint: ST/NS24/40
side joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F442250
capacity
250 mL
250 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)