Key Documents
SYNA922440
Synthware vacuum or inert gas adapter
male joint: ST/NS 24/40
Synonym(s):
Synthware A922440, glass vacuum adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
male joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A922440
Related Categories
General description
Synthware vacuum or gas inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure. Hose connection (10 mm O.D.) is at a 90 degree angle to the standard taper inner joint (24/40).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)