Key Documents
SYNA201424
Synthware reducing adapter
top joint: ST/NS 14/20, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A201424, glass reducing adapter, connecting adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
bottom joint: ST/NS24/40
joint: ST/NS top14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A201424
General description
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (24/40) is larger than the top joint (14/20).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)