Skip to Content
Merck
All Photos(2)

Documents

P5754

Sigma-Aldrich

Potassium tetraborate tetrahydrate

ReagentPlus®, ≥99.5%

Synonym(s):

Potassium biborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2B4O7 · 4H2O
CAS Number:
Molecular Weight:
305.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

product line

ReagentPlus®

Assay

≥99.5%

form

powder

SMILES string

O.O.O.O.[K+].[K+].[O-]B([O-])OB1OB(OB=O)O1

InChI

1S/B4O7.2K.4H2O/c5-1-8-3-10-4(11-3)9-2(6)7;;;;;;/h;;;4*1H2/q-2;2*+1;;;;

InChI key

FZICZLCGJZRAHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium tetraborate tetrahydrate(K2B4O7· 4H2O) is a white crystalline salt with good buffering properties. It is widely used in the field of electrochemical water splitting, disinfectant, and the oil and gas industry.

Application

Potassium tetraborate tetrahydrate can be used:

  • As an electrolyte for electrochemical water splitting reaction. The addition of K2B4O7 · 4H2O enhances the oxygen evolution activity of the catalyst.
  • To synthesize carbon foams with hierarchical pores, for supercapacitors. K2B4O7 · 4H2O helps to tune the pore structure of carbon foams.
  • To prepare green formulation for the removal of pyrite(FeS2) scales from oil and gas tubings.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ fabrication of nanostructured Au?Bi as high-efficient electrocatalyst for water splitting
Qiang Zhao, et al.
International Journal of Hydrogen Energy, 44, 14938-14944 (2019)
Development of a New Borax-Based Formulation for the Removal of Pyrite Scales
Musa Ahmed, et al.
ACS Omega, 5, 14308-14315 (2020)
Enhancement Effect of Borate Doping on the Oxygen Evolution Activity of ?-Nickel Hydroxide
Zhao Zhang, et al.
ACS Applied Nano Materials, 1, 751-758 (2018)
Thermal decomposition of potassium tetraborate tetrahydrate to anhydrous potassium tetraborate in a fluidized bed.
Sahin O, et al.
Thermochimica Acta, 440(1), 7-12 (2006)
The crystal structure of potassium tetraborate tetrahydrate.
Marezio M, et al.
Acta Crystallographica, 16(10), 975-980 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service