P5654
2,3,4,5,6-Pentafluorothiophenol
97% (GC), powder, cyclooxygenase inhibitor
Synonym(s):
PFBT, Pentafluorobenzenethiol, Pentafluorophenyl mercaptan
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6F5SH
CAS Number:
Molecular Weight:
200.13
Beilstein:
1876292
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Product Name
2,3,4,5,6-Pentafluorothiophenol, 97%
Quality Level
Assay
97%
refractive index
n20/D 1.4645 (lit.)
bp
143 °C (lit.)
mp
−24 °C (lit.)
density
1.501 g/mL at 25 °C (lit.)
SMILES string
Fc1c(F)c(F)c(S)c(F)c1F
InChI
1S/C6HF5S/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H
InChI key
UVAMFBJPMUMURT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
123.8 °F - closed cup
Flash Point(C)
51 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
James W Borchert et al.
Nature communications, 10(1), 1119-1119 (2019-03-10)
The contact resistance in organic thin-film transistors (TFTs) is the limiting factor in the development of high-frequency organic TFTs. In devices fabricated in the inverted (bottom-gate) device architecture, staggered (top-contact) organic TFTs have usually shown or are predicted to show
Rei Shiwaku et al.
Scientific reports, 8(1), 6368-6368 (2018-04-25)
Wearable sensor device technologies, which enable continuous monitoring of biological information from the human body, are promising in the fields of sports, healthcare, and medical applications. Further thinness, light weight, flexibility and low-cost are significant requirements for making the devices
Qiaoming Zhang et al.
Scientific reports, 6, 39623-39623 (2016-12-23)
Since the first demonstration, the electrolyte-gated organic field-effect transistors (EGOFETs) have immediately gained much attention for the development of cutting-edge technology and they are expected to have a strong impact in the field of (bio-)sensors. However EGOFETs directly expose their
Inés Temiño et al.
Nature communications, 11(1), 2136-2136 (2020-05-03)
Organic semiconductor materials exhibit a great potential for the realization of large-area solution-processed devices able to directly detect high-energy radiation. However, only few works investigated on the mechanism of ionizing radiation detection in this class of materials, so far. In
Panagiotis Mougkogiannis et al.
Sensors (Basel, Switzerland), 21(1) (2020-12-31)
Low power gas sensors with high sensitivity and selectivity are desired for many practical applications. Devices based on organic field effect transistors are promising because they can be fabricated at modest cost and are low power devices. Organic field effect
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service