Skip to Content
Merck
All Photos(3)

Documents

P24055

Sigma-Aldrich

1-Phenyl-1,2-ethanediol

97%

Synonym(s):

(±)-1-Phenyl-1,2-ethanediol, (±)-Phenylethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
1306723
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

272-274 °C/760 mmHg (lit.)

mp

66-68 °C (lit.)

SMILES string

OCC(O)c1ccccc1

InChI

1S/C8H10O2/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8-10H,6H2

InChI key

PWMWNFMRSKOCEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashwini C Mathpati et al.
Journal of biotechnology, 262, 1-10 (2017-09-30)
Kinetic resolution of rac-1,2-diols using the biocatalyst Burkholderia cepacia lipase (BCL) immobilized on a biodegradable binary blend support of hydroxypropyl methyl cellulose(HPMC)/polyvinyl alcohol (PVA) has been investigated. The immobilization technique improved enzyme activity significantly and it has excellent recyclability with
Li Cao et al.
Biotechnology and bioengineering, 94(3), 522-529 (2006-02-25)
Soluble epoxide hydrolase (EH) from the potato Solanum tuberosum and an evolved EH of the bacterium Agrobacterium radiobacter AD1, EchA-I219F, were purified for the enantioconvergent hydrolysis of racemic styrene oxide into the single product (R)-1-phenyl-1,2-ethanediol, which is an important intermediate
Cesar Mateo et al.
Analytical biochemistry, 314(1), 135-141 (2003-03-14)
In this paper we report the development of a novel and simple spectrophotometric assay which allows one to achieve the continuous, rapid, sensitive, and accurate determination of an epoxide hydrolase activity. This assay is based on the elaboration of a
O Spiegelstein et al.
Pharmaceutical research, 17(2), 216-221 (2000-04-06)
The purpose of this study was to evaluate the in vitro inhibitory potency of various amide analogues and derivatives of valproic acid toward human microsomal epoxide hydrolase (mEH). mEH inhibition was evaluated in human liver microsomes with 25 microM (S)-(+)-styrene
Yao Nie et al.
Organic & biomolecular chemistry, 9(11), 4070-4078 (2011-04-21)
The application of biocatalysis to the synthesis of chiral molecules is one of the greenest technologies for the replacement of chemical routes due to its environmentally benign reaction conditions and unparalleled chemo-, regio- and stereoselectivities. We have been interested in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service