Skip to Content
Merck
All Photos(1)

Key Documents

P10801

Sigma-Aldrich

Perinaphthenone

97%

Synonym(s):

1H-Benzonaphthen-1-one, 7-Perinaphthenone, Phenalenone, Phenalone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8O
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

153-156 °C (lit.)

SMILES string

O=C1C=Cc2cccc3cccc1c23

InChI

1S/C13H8O/c14-12-8-7-10-4-1-3-9-5-2-6-11(12)13(9)10/h1-8H

InChI key

WWBGWPHHLRSTFI-UHFFFAOYSA-N

Related Categories

Application

Perinaphthenone can be used:
  • In the Weitz-Scheffer epoxidation to produce the corresponding epoxides in the presence of gem-dihydroperoxide as the stoichiometric oxidant.
  • As a photosensitizer in the photodegradation of tebuconazole.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Cristina Flors et al.
Accounts of chemical research, 39(5), 293-300 (2006-05-17)
Plants defend themselves from pathogen infections or mechanical injury by a number of mechanisms, including the induced biosynthesis of antimicrobial secondary metabolites. These compounds, termed phytoalexins, represent a very economical way to counteract hazard, because the carbon and energy resources
William Hidalgo et al.
Journal of agricultural and food chemistry, 57(16), 7417-7421 (2009-07-28)
The levels of native fungitoxic perinaphthenone phytoalexins in susceptible Musa varieties (banana), which are commercially grown in large plantations, are too low to provide plants with long-lasting protection against highly pathogenic fungi. Novel strategies for plant protection are necessary to
S Opitz et al.
Planta, 216(5), 881-889 (2003-03-08)
Phenylphenalenones represent a typical group of secondary metabolites of the Haemodoraceae. Some of these phenolic compounds show organ-specific distribution within the plant. However, detailed information on cellular localisation is still lacking. To this end, confocal laser-scanning microscopy, microspectral photometry and
Mahmoud F Elsebai et al.
Natural product reports, 31(5), 628-645 (2014-04-02)
Covering up to the end of August 2013. Phenalenones are members of a unique class of natural polyketides exhibiting diverse biological potential. This is a comprehensive review of 72 phenalenones with diverse structural features originating from fungal sources. Their bioactive
Xiaopeng Chen et al.
Chemical communications (Cambridge, England), 47(9), 2628-2630 (2011-01-15)
Phenalenone derivatives were efficiently constructed from 1,8-diiodonaphthalene and tertiary propynols via a one-pot domino reaction which eventually included Pd-catalyzed Sonogoshira coupling, Pd-catalyzed allylic oxidation and Pd-catalyzed C(sp(2))-H activation. Moreover, the synthesized phenalenone derivative presented a practical application as a fluorescent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service