OCNQSS450
Cd-based Core/Shell Quantum Dots with Streptavidin
fluorescence λem 450 nm, 1 μM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
43211915
NACRES:
NA.23
Recommended Products
concentration
1 μM
fluorescence
λem 450 nm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Streptavidin has high binding affinity to biotin or biotin-containing molecules such as proteins, peptides, polysaccharides, oligonucleotides. Streptavidin functionalized quantum dots offer an simple and universal method to conjugate those biotinylated molecules to quantum dots for downstream applications such as flow cytometry, Westen blot, microplate assay, fluorescent imaging with microscope for slide-based assay , microarray assay, bead-based assay, fluorescent sensing.The advantages of using quantum dots as fluorophores include high brightness, narrow and symmetric emission, resistant to photobleaching, etc. These unique features make quantum dots ideal optical probes for highly sensitive multiplexing applications.
Legal Information
Product of Ocean Nanotech, LLC.
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service