Skip to Content
Merck
All Photos(1)

Key Documents

NCXSCPH100

Sigma-Aldrich

Silver Nanocubes

100 nm, NanoXact, 1 mg/mL in ethanol, PVP 55 kDa (Polymer)

Synonym(s):

Silver Nano-cubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanocubes

concentration

1 mg/mL in ethanol

particle size

100 nm

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our high quality silver nanocubes offer uniform flat surfaces, which makes them useful for sensors and imaging applications. These nanocubes possess narrow size distribution and exhibit surface plasmon resonance (SPR) with multiple plasmonic modes in the visible and near infrared regions. The distinctive morphology and homogeneity of these nanocubes makes them ideal for use in: surface-enhanced Raman scattering (SERS), plasmonic based catalysis, sensors, photovoltaics, and other energy harvesting devices.
Silver nanocubes (100 nm, NanoXact, 1 mg/mL in ethanol, PVP 55 kDa (polymer)) can be potentially used for a variety of applications such as surface enhanced Raman scattering (SERS) substrate, MoS2 photodetector, light-emitting diodes (LEDs), and localized surface plasmon resonance (LSPR).

Storage and Stability

DO NOT FREEZE. Store at 2-8C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of silver nanocubes as a SERS substrate for the determination of pesticide paraoxon and thiram
Wang B, et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 121, 63-69 (2014)
Localized surface plasmon resonance spectroscopy of single silver nanocubes
Sherry LJ, et al.
Nano Letters, 5(10), 2034-2038 (2005)
Tailoring of Silver Nanocubes with Optimized Localized Surface Plasmon in a Gap Mode for a Flexible MoS2 Photodetector
Sun B, et al.
Advances in Functional Materials, 29(26), 1900541-1900541 (2019)
High-performance solution-processed white organic light-emitting diodes based on silica-coated silver nanocubes
Deng L, et al.
Optics Letters, 44(4), 983-986 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service