Skip to Content
Merck
All Photos(3)

Key Documents

N32601

Sigma-Aldrich

Norcamphor

98%

Synonym(s):

2-Norbornanone, Bicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O
CAS Number:
Molecular Weight:
110.15
Beilstein:
1209657
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

168-172 °C (lit.)

mp

93-96 °C (lit.)

SMILES string

O=C1C[C@@H]2CC[C@H]1C2

InChI

1S/C7H10O/c8-7-4-5-1-2-6(7)3-5/h5-6H,1-4H2/t5-,6+/m1/s1

InChI key

KPMKEVXVVHNIEY-RITPCOANSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J Contzen et al.
Biochemistry, 37(13), 4317-4324 (1998-04-29)
Step-scan time-resolved Fourier transform infrared spectroscopy with a time resolution of 5 micros was applied to the carbon monoxide complex of cytochrome P-450cam (CYP101) to study the bimolecular ligand-rebinding process after flash photolysis. Spectral changes in the CO ligand stretch
M B Bass et al.
Proteins, 13(1), 26-37 (1992-05-01)
While cytochrome P-450cam catalyzes the hydroxylation of camphor to 5-exo-hydroxycamphor with 100% stereospecificity, norcamphor is hydroxylated by this enzyme yielding 45% 5-exo-, 47% 6-exo-, and 8% 3-exo-hydroxynorcamphor (Atkins, W.M., Sligar, S.G., J. Am. Chem. Soc. 109:3754-3760, 1987). The present study
Christiane Jung et al.
European journal of biochemistry, 269(12), 2989-2996 (2002-06-20)
The high-pressure stopped-flow technique is applied to study the CO binding in cytochrome P450cam (P450cam) bound with homologous substrates (1R-camphor, camphane, norcamphor and norbornane) and in the substrate-free protein. The activation volume DeltaV # of the CO on-rate is positive
J R Collins et al.
The Journal of biological chemistry, 263(7), 3164-3170 (1988-03-05)
The hydroxylations of d-camphor, norcamphor, pericyclocamphanone, and 5,5-difluorocamphor by cytochrome P-450cam have been examined using theoretical methods to identify and characterize properties which determine product specificity. Experimental results indicate that each molecule is hydroxylated with quite different regio-specificity when metabolized
Megan C Thielges et al.
The journal of physical chemistry. A, 115(34), 9714-9723 (2011-06-09)
Progress in the field of 2D IR vibrational spectroscopy has been bolstered by the production of intense mid-IR laser pulses. As higher-energy pulses are employed, a concomitant increase occurs in the likelihood of fifth-order contributions to the 2D IR spectra.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service