All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H11NO
CAS Number:
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥98%
form
solid
storage temp.
−20°C
SMILES string
COc1cc(N)cc2ccccc12
InChI
1S/C11H11NO/c1-13-11-7-9(12)6-8-4-2-3-5-10(8)11/h2-7H,12H2,1H3
InChI key
SFKZPTYRENGBTJ-UHFFFAOYSA-N
Related Categories
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Heidi A Kluess et al.
Medicine, 98(13), e14982-e14982 (2019-03-29)
The purpose was to investigate changes in neuropeptide Y (NPY) protein and dipeptidyl peptidase IV (DPP-IV) activity in the plasma and saliva in normally cycling women and women after menopause. We recruited 7 cycling women and 7 postmenopausal women for
C N Kennett et al.
Journal of periodontal research, 29(3), 203-213 (1994-05-01)
Cathepsin B activity was demonstrated histochemically in unfixed cryostat sections of inflamed human gingiva using the 2-methoxy-4-naphthylamide (MNA) substrates Z-Val-Lys-Lys-Arg-MNA and Z-Ala-Arg-Arg-MNA with a post-azo-coupling technique. Enzyme localisation was confirmed by immunocytochemistry with polyclonal sheep anti-human cathepsin B. In both
B Ulbricht et al.
Biological chemistry Hoppe-Seyler, 376(7), 407-414 (1995-07-01)
The cysteine proteases cathepsin B and cathepsin L are very likely involved in invasive processes of normal and malignant cells, they become relevant for a number of diseases and are possibly prognostic markers for the outcome of human lung cancer.
Epidermal aminopeptidase activity and metabolism as observed in an organized HaCaT cell sheet model.
I Steinsträsser et al.
Journal of pharmaceutical sciences, 86(3), 378-383 (1997-03-01)
Metabolism studies on organized HaCaT keratinocyte cell sheets are reported. Cells were grown on porous membranes to form organized cell sheets of several cell layers, which were considered as a model of viable epidermis. Metabolism was studied by reflection kinetics
S H Randell et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 33(7), 677-686 (1985-07-01)
Dipeptidyl peptidase II (DPP II) in normal rat lung was evaluated by the enzymes' ability to hydrolyze Lys-Ala or Lys-Pro derivatives of 4-methoxy-2-naphthylamine (MNA). For visualization of this activity, the liberated MNA was coupled with fast blue B for light
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service