M86502
Methyl trimethylacetate
99%
Synonym(s):
Methyl pivalate, Methyl 2,2-dimethylpropionate, Methyl trimethylacetate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(CH3)3CCOOCH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1744141
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.390 (lit.)
bp
101 °C (lit.)
density
0.873 g/mL at 25 °C (lit.)
SMILES string
COC(=O)C(C)(C)C
InChI
1S/C6H12O2/c1-6(2,3)5(7)8-4/h1-4H3
InChI key
CNMFHDIDIMZHKY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
42.8 °F - closed cup
Flash Point(C)
6 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service