Skip to Content
Merck
All Photos(1)

Key Documents

M81101

Sigma-Aldrich

mono-Methyl hydrogen succinate

95%

Synonym(s):

Monomethyl succinate, Succinic acid monomethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2CH2COOH
CAS Number:
Molecular Weight:
132.11
Beilstein:
1722669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

bp

151 °C/20 mmHg (lit.)

mp

54-57 °C (lit.)

SMILES string

COC(=O)CCC(O)=O

InChI

1S/C5H8O4/c1-9-5(8)3-2-4(6)7/h2-3H2,1H3,(H,6,7)

InChI key

JDRMYOQETPMYQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Lajoix et al.
Diabetes, 50(6), 1311-1323 (2001-05-26)
Evidence is presented showing that a neuronal isoform of nitric oxide synthase (NOS) is expressed in rat pancreatic islets and INS-1 cells. Sequencing of the coding region indicated a 99.8% homology with rat neuronal NOS (nNOS) with four mutations, three
L Ladrière et al.
Annals of nutrition & metabolism, 41(2), 118-125 (1997-01-01)
Rats were fasted for 48 h, but infused with either NaCl or the sodium salt of monoethyl succinic acid (EMS), both delivered at a rate of 80 mumol/g body weight per day. The infusion of EMS, as compared to NaCl
S A Hinke et al.
British journal of pharmacology, 150(8), 1031-1043 (2007-03-07)
Two mechanisms have been proposed to explain the insulin-sensitising properties of metformin in peripheral tissues: (a) inhibition of electron transport chain complex I, and (b) activation of the AMP activated protein kinase (AMPK). However the relationship between these mechanisms and
K Capito et al.
Journal of molecular endocrinology, 17(2), 101-107 (1996-10-01)
The involvement of phosphatidylcholine (PC) hydrolysis in the regulation of insulin secretion was studied in mouse pancreatic islets prelabelled with [3H]choline. Phospholipase C (PLC) and phospholipase D (PLD) activities were demonstrated and also that of an enzyme that removes both
Y P Zhou et al.
The American journal of physiology, 270(6 Pt 1), E988-E994 (1996-06-01)
Fasting inhibits glucose-induced insulin secretion. We investigated the role of a glucose fatty acid cycle for such inhibition and its molecular basis in pancreatic islets from 48-h fasted rats. The fasting-impaired insulin response to 27 mM glucose was restored by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service