Skip to Content
Merck
All Photos(3)

Key Documents

M80903

Sigma-Aldrich

α-Methylstyrene

99%, contains 15 ppm p-tert-butylcatechol as inhibitor

Synonym(s):

2-Phenylpropene, Isopropenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)=CH2
CAS Number:
Molecular Weight:
118.18
Beilstein:
969405
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

4.1 (vs air)

Quality Level

vapor pressure

2.1 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

1065 °F

contains

15 ppm p-tert-butylcatechol as inhibitor

expl. lim.

6.1 %

refractive index

n20/D 1.538 (lit.)

bp

165-169 °C (lit.)

mp

−24 °C (lit.)

density

0.909 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)c1ccccc1

InChI

1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3

InChI key

XYLMUPLGERFSHI-UHFFFAOYSA-N

General description

-Methylstyrene is an aromatic alkyl-substituted styrene monomer with a relatively high boiling point. It is widely used as a monomer to synthesize thermoplastic composite resins applied in adhesives, and coatings that require high-temperature stability. It can also participate as a precursor in regioselective and diastereoselective synthesis of natural products, pharmaceuticals, and other structurally diverse compounds.

Application

α-Methylstyrene can be used:

  • As a monomer to prepare “Phene” monomer for photopolymerizable dental resin composites to reduce their polymerization shrinkage. α-Methylstyrene has a low double bond concentration suitable for the preparation of low volumetric shrinkage and shrinkage stress resilient resin composites.
  • As a monomer to prepare poly(α-methylstyrene) polymer which can be used as an additive to fabricate organic thin film transistors. This polymer promotes the growth of uniformly aligned semiconductor crystals with significantly improved orientation.
  • As a precursor to synthesize 3,4-dihydropyrans with therapeutic potential.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic Diastereoselective Hetero-Diels?Alder Reaction of ?-Haloacroleins with Alkenes: Construction of 3,4-Dihydropyran
Lei Zeng, et al.
Organic Letters, 24, 2115-2119 (2022)
Ting Luo et al.
iScience, 23(11), 101717-101717 (2020-11-17)
Resin embedding is widely used to dissect the fine structure of bio-tissue with electron and optical microscopy. However, it is difficult to embed large-volume tissues with resin. Here, we modified the formula of LR-White resin to prevent the sample cracking
A N Ilialetdinov et al.
Mikrobiologiia, 52(4), 609-614 (1983-07-01)
Two bacterial strains assimilating alpha-methylsterene as a sole carbon and energy source were isolated from the sewage of an industrial plant producing synthetic rubber. The morphological, cultural and physiologo-biochemical properties of the strains were studied and their taxonomic position was
[Kallikrein-kinin system indices in neurocirculatory dystonia patients who work in synthetic rubber manufacture].
N I Al'berton et al.
Gigiena truda i professional'nye zabolevaniia, (12)(12), 23-26 (1981-12-01)
The effect of adding a new monomer ?Phene? on the polymerization shrinkage reduction of a dental resin composite
Jingwei He, et al.
Dental Materials Journal, 35, 627-635 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service