Skip to Content
Merck
All Photos(1)

Key Documents

M3394

Sigma-Aldrich

6-Methyl-2,4-heptanedione

≥98%

Synonym(s):

Isovalerylacetone, NSC 42238, NSC 42240, NSC 46469, NSC 94

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH2COCH3
CAS Number:
Molecular Weight:
142.20
Beilstein:
1751122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

color

colorless to faint yellow

density

0.92 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CC(=O)CC(C)=O

InChI

1S/C8H14O2/c1-6(2)4-8(10)5-7(3)9/h6H,4-5H2,1-3H3

InChI key

IGMOYJSFRIASIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant involved in:
  • Cyclocondensation and deacetylation of dibromo(phenylsulfonyl)propene derivatives for synthesis of disubstituted furans
  • Synthesis of dual activity effectors of angiotensin II type 1 receptors and peroxisome proliferator-activated receptor-γ
  • Addition reactions with 1-alkynes
  • Knoevenagel condensation
  • Oxidative free radical reactions with 2-amino-1,4-benzoquinones
  • Mild oxidative cleavage for synthesis of carboxylic acids

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iwona Łakomska et al.
Materials (Basel, Switzerland), 13(23) (2020-12-02)
Herein, we present dicarboxylate platinum(II) complexes of the general formula [Pt(mal)(DMSO)(L)] and [Pt(CBDC)(DMSO)(L)], where L is dbtp 5,7-ditertbutyl-1,2,4-triazolo[1,5-a]pyrimidine) or ibmtp (7-isobutyl-5-methyl-1,2,4- triazolo[1,5-a]pyrimidine), as prospective prodrugs. The platinum(II) complexes were synthesized in a one-pot reaction between cis-[PtCl2(DMSO)2], silver malonate or silver
Marzena Fandzloch et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 25(1), 109-124 (2019-11-20)
Six novel ruthenium(III) complexes of general formula [RuCl3(L)3] (1,3,5) and [RuCl3(H2O)(L)2] (2,4,6), where L stands for three different triazolopyrimidine-derived ligands, are reported. The compounds have been structurally characterized (IR, EPR, SCXRD), and their magnetic moments have been determined. The single-crystal
Marzena Fandzloch et al.
Journal of inorganic biochemistry, 210, 111072-111072 (2020-06-21)
Five novel rhodium(II) complexes of general formula [Rh2(μ-OOCCH3)4L2], where L is a triazolopyrimidine derivative, in particular dimethyl-1,2,4-triazolo[1,5-a]pyrimidine (dmtp) for (1), 5,7-diethyl-1,2,4-triazolo[1,5-a]pyrimidine (detp) for (2), 7-isobutyl-5-methyl-1,2,4-triazolo[1,5-a]pyrimidine (ibmtp) for (3), 7-hydroxy-5-methyl-1,2,4-triazolo[1,5-a]pyrimidine (HmtpO) for (4) and 5,7-ditertbutyl-1,2,4-triazolo[1,5-a]pyrimidine (dbtp) for (5) are reported. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service