Skip to Content
Merck
All Photos(1)

Key Documents

L030080

Sigma-Aldrich

ETT Activator

0.25 M, configured for PerkinElmer, configured for Polygen, configured for MerMade, configured for ABI 3900

Synonym(s):

5-(Ethylthio)-1H-tetrazole, 5-(Ethylthio)tetrazole solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

concentration

32-35 g/L (ETT content, titration)

impurities

≤30 ppm water content (Karl Fischer)

color

clear

compatibility

configured for ABI 3900
configured for MerMade
configured for PerkinElmer
configured for Polygen

SMILES string

CCSc1nnn[nH]1

InChI

1S/C3H6N4S/c1-2-8-3-4-6-7-5-3/h2H2,1H3,(H,4,5,6,7)

InChI key

GONFBOIJNUKKST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetonitrile / 5-(Ethylthio)-1H-Tetrazole 100 / 3 (v/w)

Other Notes

Activator solution, containing 5-(Ethylthio)-1H- tetrazole (ETT) in acetonitrile, are mixed with solutions of phosphoramidites during the extension step.
The activator reacts with the amidite group to form a highly reactive intermediate. The intermediate then forms an internucleotide bond with the detritylated 5′hydroxyl group of the growing oligonucleotide chain.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service