KIM3450599
Kimble® melting point capillary
case of 20 × 100 ea (2000 total), borosilicate glass, O.D. × L 1.5-1.8 mm × 90 mm, sealed at one end
Synonym(s):
34505-99, Capillary Melting Point, Capillary Tubes, Melting Point Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
borosilicate glass
packaging
case of 20 × 100 ea (2000 total)
O.D. × L
1.5-1.8 mm × 90 mm
General description
KIMAX® melting point capillary tubes are used to determine the point at which a solid sample turns to liquid.
Features and Benefits
Melting point capillary tubes are used to determine the point at which a solid sample turns to liquid.
- Tube length insures that the open end will extend well above the bath′s surface to prevent the bath liquid from entering the tube
- Packaged with 100 tubes per vial and 20 vials per case
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service