Skip to Content
Merck
All Photos(1)

Key Documents

H2201

Sigma-Aldrich

1,7-Heptanediol

95%

Synonym(s):

Heptamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)7OH
CAS Number:
Molecular Weight:
132.20
Beilstein:
1633482
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

liquid

refractive index

n20/D 1.455 (lit.)

bp

259 °C (lit.)

mp

17-19 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCCO

InChI

1S/C7H16O2/c8-6-4-2-1-3-5-7-9/h8-9H,1-7H2

InChI key

SXCBDZAEHILGLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Stortz et al.
BMC biology, 18(1), 59-59 (2020-06-04)
Functional compartmentalization has emerged as an important factor modulating the kinetics and specificity of biochemical reactions in the nucleus, including those involved in transcriptional regulation. The glucocorticoid receptor (GR) is a ligand-activated transcription factor that translocates to the nucleus upon
S V Vinogradov et al.
Biochemical and biophysical research communications, 203(2), 959-966 (1994-09-15)
Antisense oligonucleotides were modified by their ends with hydrophobic substituents which permitted enhancing their activity. The effect of hydrophobized oligonucleotides on reproduction of Herpes Simplex Virus type 1 in Vero cells was studied. Two types of oligonucleotides were used: a
Fabrian Brenz et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(2), 307-319 (2016-11-29)
Polyesters (PESs) are gaining more importance on the food contact material (FCM) market and the variety of properties and applications is expected to be wide. In order to acquire the desired properties manufacturers can combine several FCM-approved polyvalent carboxylic acids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service