Skip to Content
Merck
All Photos(2)

Documents

H17082

Sigma-Aldrich

2-Hydrazinopyridine

97%

Synonym(s):

2-Pyridylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
Beilstein:
109984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

90-92 °C/1 mmHg (lit.)

mp

41-44 °C (lit.)

storage temp.

2-8°C

SMILES string

NNc1ccccn1

InChI

1S/C5H7N3/c6-8-5-3-1-2-4-7-5/h1-4H,6H2,(H,7,8)

InChI key

NWELCUKYUCBVKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Enhanced Metal Removal: Utilizing Schiff base functionalized dialdehyde starch, derived from 2-hydrazinopyridine, demonstrated significant potential in enhancing the removal of Cu(II) from solutions. The study included preparation methodologies, performance evaluations, and DFT calculations, showcasing its efficacy in water treatment technologies (Liang et al., 2024).
  • Dual Sensing Probe Development: A novel dicyanisophorone-based probe, incorporating 2-hydrazinopyridine, was developed for the dual sensing of Zn(2+) and Cd(2+) via near-infrared fluorescence. This advancement aids in the detection and analysis of heavy metals in various environmental and biological samples (Yan et al., 2023).
  • Active Site Analysis in Lysyl Oxidase: The study provided insights into the spatial arrangement of active site components in Lysyl Oxidase-like 2, including the role of 2-hydrazinopyridine, which is critical for understanding the enzyme′s mechanism and potential therapeutic applications (Meier et al., 2022).
  • Structural Analysis of Lysyl Oxidase: Research focused on the predicted 3D structure of the amine oxidase domain of Lysyl Oxidase-Like 2, exploring the interaction dynamics facilitated by 2-hydrazinopyridine. This contributes significantly to the field of molecular biology and enzyme function analysis (Meier et al., 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Knowles et al.
Journal of neural transmission (Vienna, Austria : 1996), 114(6), 743-746 (2007-04-05)
Tranylcypromine (TCP), an amphetamine, is a reversible inhibitor of copper-containing amine oxidases. We have solved the structure of the complex of TCP with the amine oxidase from E. coli (ECAO) and shown that only the (+)-enantiomer of TCP binds. Kinetic
Christian R P Kurtis et al.
Journal of neural transmission (Vienna, Austria : 1996), 118(7), 1043-1053 (2011-05-07)
Copper amine oxidases are important for the metabolism of a range of biogenic amines. Here, we focus on substrate specificity in the E. coli copper amine oxidase (ECAO) and specifically the role of Tyr 381. This residue, and its equivalent
Sudath Hapuarachchige et al.
Journal of the American Chemical Society, 133(17), 6780-6790 (2011-04-09)
A new class of fluorescent triazaborolopyridinium compounds was synthesized from hydrazones of 2-hydrazinylpyridine (HPY) and evaluated as potential dyes for live-cell imaging applications. The HPY dyes are small, their absorption/emission properties are tunable through variation of pyridyl or hydrazone substituents
Minae Mure et al.
Biochemistry, 44(5), 1583-1594 (2005-02-03)
Adduct I (lambda(max) at approximately 430 nm) formed in the reaction of 2-hydrazinopyridine (2HP) and the TPQ cofactor of wild-type Escherichia coli copper amine oxidase (WT-ECAO) is stable at neutral pH, 25 degrees C, but slowly converts to another spectroscopically
E S H El Ashry et al.
Nucleosides, nucleotides & nucleic acids, 23(3), 567-580 (2004-04-29)
Reaction of 2-hydrazinopyridine (1) with D-xylose, D-galactose, D-glucose and D-fructose afforded the corresponding hydrazones mainly in the acyclic forms 2, 3, 6 and 11 with minor amounts of the cyclic structures. Oxidative cyclization of the hydrazones with bromine in methanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service