Skip to Content
Merck
All Photos(1)

Key Documents

H1604

Sigma-Aldrich

2,2,3,3,4,4,4-Heptafluoro-1-butanol

98%

Synonym(s):

Perfluoropropyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2CH2OH
CAS Number:
Molecular Weight:
200.05
Beilstein:
1761907
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.3 (lit.)

bp

96-97 °C (lit.)

density

1.6 g/mL at 25 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4H3F7O/c5-2(6,1-12)3(7,8)4(9,10)11/h12H,1H2

InChI key

WXJFKAZDSQLPBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,3,3,4,4,4-Heptafluoro-1-butanol can be used to synthesize poly(2-hydroxyethyl vinyl ether)-block-poly(2-(2,2,3,3,4,4,4-heptafluorobutoxy)ethyl vinyl ether) (poly(HOVE-b-HFBOVE), a fluorinated amphiphilic block copolymer.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dai Long Vu et al.
Food chemistry, 279, 12-19 (2019-01-07)
Selenium (Se) is an indispensable microelement in our diet and health issues resulting from deficiencies are well documented. Se-containing food supplements are available on the market including Se-enriched Chlorella vulgaris (Se-Chlorella) which accumulates Se in the form of Se-amino acids
Fluorinated amphiphilic vinyl ether block copolymers: Synthesis and characteristics of their micelles in water.
Matsumoto K, et al.
Macromolecules, 37(6), 2256-2267 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service