Skip to Content
Merck
All Photos(2)

Documents

GF95374222

Lead

foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%

Synonym(s):

Lead, PB000280

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pb
CAS Number:
Molecular Weight:
207.20
MDL number:
UNSPSC Code:
12141723
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 953-742-22

resistivity

20.65 μΩ-cm

bp

1740 °C (lit.)

mp

327.4 °C (lit.)

SMILES string

[Pb]

InChI

1S/Pb

InChI key

WABPQHHGFIMREM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina Lopes Devito et al.
Journal of applied oral science : revista FOB, 12(4), 290-293 (2004-12-01)
All materials added to teeth should present an adequate radiopacity to allow the detection of secondary caries. Usually, in extensive cavities, base materials like calcium hydroxide cement are used for the purpose of protecting the pulp. In an attempt to
Leonard J S Tsuji et al.
Journal (Canadian Dental Association), 71(1), 35-38 (2005-01-15)
The lead content of the foil backing of 4 types of intraoral film commonly used by dentists was 69% to 85%. An environmental issue exists because these foils are typically thrown out with regular refuse, even though recycling programs exist.
Steven D Fallon et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 64(2), 223-229 (2006-01-18)
The purpose of this study was to determine if condylar morphology can be accurately depicted on the panoramic radiograph and if variations in condylar anatomy affect the radiographic appearance of the condyle. The shape and angulation of the condyle were
Antonio Lopez Medina et al.
Physics in medicine and biology, 49(1), 17-32 (2004-02-20)
Two dosimetry protocols based on absorbed dose to water have recently been implemented: TG-51 and TRS-398. These protocols use different beam-quality indices: %dd(10)x and TPR20,10. The effect of electron contamination in measurements of %dd(10)x has been proposed as a disadvantage
H Fujita et al.
The British journal of radiology, 82(978), 504-508 (2009-01-21)
It is useful to utilise a computed radiography system with a combination of storage phosphor plates and therapy cassettes for portal images. However, the therapy cassette has some disadvantages. The objective of the present study was to determine whether the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service