Skip to Content
Merck
All Photos(2)

Key Documents

GF68941390

Cadmium

foil, 50x50mm, thickness 1.0mm, as rolled, 99.95%

Synonym(s):

Cadmium, CD000272

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41
Beilstein:
8137359
MDL number:
UNSPSC Code:
12141707
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 689-413-90

resistivity

7.27 μΩ-cm, 22°C

size × thickness

50x50 mm × 1.0 mm

bp

765 °C (lit.)

mp

320.9 °C (lit.)

density

8.65 g/mL at 25 °C (lit.)

SMILES string

[Cd]

InChI

1S/Cd

InChI key

BDOSMKKIYDKNTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uroš Cvelbar et al.
Nanotechnology, 19(40), 405605-405605 (2008-10-08)
Plasma-assisted synthesis of nanostructures is one of the most precise and effective approaches used in nanodevice fabrication. Here we report on the innovative approach of synthesizing nanostructured cadmium oxide films on Cd substrates using a reactive oxygen plasma-based process. Under
Jungsoo Kim et al.
Environmental monitoring and assessment, 186(4), 2321-2327 (2014-01-11)
In birds, concentrations of heavy metal contaminants were associated with their diet and environmental quality. This study presents concentrations of lead and cadmium in feathers and the relationship between concentrations in the feather and stomach content of three species of
S E Dal-Medico et al.
Ecotoxicology and environmental safety, 104, 168-174 (2014-04-01)
A drastic amphibian decline has been observed worldwide, which can be attributed (among other factors) to exposure to pollutants. Considering that cadmium corresponds to the most rapidly increasing trace metal in the environment, the aim of this work was to
Sandra Götze et al.
Aquatic toxicology (Amsterdam, Netherlands), 149, 65-82 (2014-02-28)
Increased anthropogenic emission of CO2 changes the carbonate chemistry and decreases the pH of the ocean. This can affect the speciation and the bioavailability of metals in polluted habitats such as estuaries. However, the effects of acidification on metal accumulation
Peerapat Kosolsaksakul et al.
Environmental pollution (Barking, Essex : 1987), 187, 153-161 (2014-02-08)
The Mae Tao watershed, northwest Thailand, has become contaminated with cadmium (Cd) as a result of zinc ore extraction (Padaeng deposit) in the nearby Thanon-Thongchai mountains. Consumption of contaminated rice has led to documented human health impacts. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service