GF68499747
Zinc
foil, 1m coil, thickness 0.175mm, as rolled, 99.95+%
Synonym(s):
Zinc, ZN000290
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
1 mmHg ( 487 °C)
Assay
99.95%
form
foil
manufacturer/tradename
Goodfellow 684-997-47
resistivity
5.8 μΩ-cm, 20°C
diam. × thickness
1 m × 0.175 mm
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Conghua Lu et al.
Chemical communications (Cambridge, England), (33)(33), 3551-3553 (2006-08-22)
Large-scale, ultralong ZnO nanowire and nanobelt arrays with honeycomb-like micropatterns have been fabricated by hydrothermal oxidation of zinc foil in aqueous alkaline (NH4)2S2O8 solutions.
A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)
S Rondot et al.
Science (New York, N.Y.), 263(5154), 1739-1741 (1994-03-25)
An x-ray projection microscope equipped with a charge-coupled device camera allows direct observation of zinc (Zn(2+)) ions diffusing in aqueous hydrochloric acid solution during the corrosion of zinc foil and pellets. Time series of microradiographic images with a lateral resolution
Chenglin Yan et al.
The journal of physical chemistry. B, 110(51), 25850-25855 (2006-12-22)
Vertically aligned ZnO nanorods with uniform diameter and length have been synthesized on a zinc foil substrate with ammonium persulfate as oxidant via a facile, larger scale production and inexpensively synthesized method without any templates or additives. SEM and XRD
Tandra Ghoshal et al.
Journal of nanoscience and nanotechnology, 9(9), 5586-5591 (2009-11-26)
Nanowire arrays of zinc oxide were synthesized on zinc foil by a simple thermal evaporation process. Morphologies and sizes of the synthesized nanostructures were varied by varying the reaction time and the surface roughness of the substrate. Self-catalytic liquid-solid mechanism
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service