Skip to Content
Merck
All Photos(2)

Documents

GF63377017

Lanthanum

rod, 100mm, diameter 6.35mm, cast, 99%

Synonym(s):

Lanthanum, LA007910

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
La
CAS Number:
Molecular Weight:
138.91
MDL number:
UNSPSC Code:
12141607
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

rod

manufacturer/tradename

Goodfellow 633-770-17

resistivity

54 μΩ-cm

L × diam.

100 mm × 6.35 mm

bp

3464 °C (lit.)

mp

920 °C (lit.)

density

6.19 g/mL at 25 °C (lit.)

SMILES string

[La]

InChI

1S/La

InChI key

FZLIPJUXYLNCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monique P Curran et al.
Drugs, 69(16), 2329-2349 (2009-10-27)
Orally administered lanthanum carbonate (Fosrenol) dissociates in the acid environment of the upper gastrointestinal tract to release the cation lanthanum, which then binds dietary phosphate. Lanthanum carbonate was effective in reducing levels of serum phosphate and serum calcium x phosphate
Yongsheng Yang et al.
Molecular pharmaceutics, 10(2), 544-550 (2012-12-20)
A conventional human pharmacokinetic (PK) in vivo study is often considered as the "gold standard" to determine bioequivalence (BE) of drug products. However, this BE approach is not always applicable to the products not intended to be delivered into the
Stephen J P Damment et al.
Clinical pharmacokinetics, 47(9), 553-563 (2008-08-14)
Lanthanum carbonate is considered to be the most potent of a new generation of noncalcium phosphate binders used to treat hyperphosphataemia in chronic kidney disease (CKD), a condition associated with progressive bone and cardiovascular pathology and a markedly elevated risk
Tilman B Drüeke
Seminars in dialysis, 20(4), 329-332 (2007-07-20)
Controlling serum phosphorus levels in patients with renal failure is critical. The use of oral phosphate-binding agents is universal for patients with end-stage kidney disease to reduce phosphate absorption. The therapeutic goal is to reduce serum phosphorus levels without disturbing
M Cozzolino et al.
The International journal of artificial organs, 30(4), 293-300 (2007-05-24)
Phosphate overload is a dramatic consequence in end-stage renal disease (ESRD) patients. Recent studies have well documented that abnormalities in mineral and bone metabolism in these patients are associated with increased cardiovascular morbidity and mortality. Elevated serum phosphate and calcium-phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service