GF63273731
Terbium
rod, 100mm, diameter 6.35mm, cast, 99%
Synonym(s):
Terbium, TB007910
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Tb
CAS Number:
Molecular Weight:
158.93
MDL number:
UNSPSC Code:
12141614
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99%
form
rod
manufacturer/tradename
Goodfellow 632-737-31
resistivity
116 μΩ-cm, 20°C
L × diam.
100 mm × 6.35 mm
bp
3230 °C (lit.)
mp
1356 °C (lit.)
density
8.234 g/mL at 25 °C (lit.)
SMILES string
[Tb]
InChI
1S/Tb
InChI key
GZCRRIHWUXGPOV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhi Shi et al.
Cancer research, 75(1), 147-158 (2014-11-02)
Cables1 is a candidate tumor suppressor that negatively regulates cell growth by inhibiting cyclin-dependent kinases. Cables1 expression is lost frequently in human cancer but little is known about its regulation. Here, we report that Cables1 levels are controlled by a
Dinari A Harris et al.
Current protocols in nucleic acid chemistry, Chapter 6, Unit 6-Unit 6 (2008-04-23)
The function of an RNA molecule is determined by its overall secondary and tertiary structure. The tertiary structure is facilitated and stabilized by the interaction with metal ions. The current chapter offers a detailed protocol on the use of the
Lei Rao et al.
International journal of food microbiology, 205, 73-80 (2015-04-19)
The objective of this study was to investigate the inactivation of the Bacillus subtilis spores by high pressure CO2 combined with high temperature (HPCD+HT) and to analyze the clumping effect of the spores on their HPCD+HT resistance. The spores of
Cédric Brulé et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(12), 5148-5162 (2014-09-04)
Biased agonism by G-protein-coupled receptor ligands has opened up strategies for targeted physiological or therapeutic actions. We hypothesized that urotensin II (UII)-derived peptides displayed unexpected physiological effects because of such biased signaling on the UII human urotensin (hUT) receptor. We
Margaret L Warner et al.
Molecular pharmacology, 87(3), 410-420 (2014-12-17)
Secreted R-spondin proteins (RSPOs1-4) function as adult stem cell growth factors by potentiating Wnt signaling. Simultaneous binding of distinct regions of the RSPO Fu1-Fu2 domain module to the extracellular domains (ECDs) of the LGR4 G protein-coupled receptor and the ZNRF3
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service