Skip to Content
Merck
All Photos(1)

Key Documents

GF60948658

Ruthenium

Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%

Synonym(s):

Ruthenium, RU004200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ru
CAS Number:
Molecular Weight:
101.07
MDL number:
UNSPSC Code:
12141739
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9%

manufacturer/tradename

Goodfellow 609-486-58

resistivity

7.1 μΩ-cm, 0°C

bp

3900 °C (lit.)

mp

2310 °C (lit.)

density

12.45 g/cm3 (lit.)

SMILES string

[Ru]

InChI

1S/Ru

InChI key

KJTLSVCANCCWHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole L Fry et al.
Accounts of chemical research, 44(4), 289-298 (2011-03-03)
Nitric oxide (NO) can induce apoptosis (programmed cell death) at micromolar or higher doses. Although cell death via NO-induced apoptosis has been studied quite extensively, the targeted delivery of such doses of NO to infected or malignant tissues has not
Lutz Ackermann et al.
Topics in current chemistry, 292, 211-229 (2010-01-01)
Stoichiometric cycloruthenation reactions of substrates containing Lewis-basic functionalities set the stage for efficient ruthenium-catalyzed C-H bond functionalization reactions. Thereby, selective addition reactions of C-H bonds across alkenes or alkynes enabled atom-economical synthesis of substituted arenes. More recently, ruthenium-catalyzed direct arylation
Gregory S Smith et al.
Dalton transactions (Cambridge, England : 2003), 40(41), 10793-10800 (2011-08-23)
The introduction of multifunctionalities for tumour targeting is becoming a popular strategy toward the development of new therapeutic agents. In particular, the multifaceted potential of ruthenium(II)-arene complexes show great promise as chemotherapeutics. An ever-increasing number of papers dealing with the
Juliano Andreoli Miyake et al.
Anticancer research, 34(4), 1901-1911 (2014-04-03)
Gliomas are primary brain tumours. Gamma-linolenic acid (GLA) exerts anti-proliferative effects. Several ruthenium-containing complexes have antiproliferative effects and can be used as adjuvant therapies in cisplatin-resistant cancer. The present study reports on the anti-proliferative properties and effects on tumour morphology
Christian G Hartinger et al.
Current topics in medicinal chemistry, 11(21), 2688-2702 (2011-11-02)
Polynuclear compounds are a relatively new and successful approach in metal-based cancer chemotherapy as typified by the trinuclear Pt compound BBR3464 which was evaluated in clinical trials. In this review, we discuss newer developments of polynuclear ruthenium, osmium and gold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service