Skip to Content
Merck
All Photos(2)

Key Documents

GF37370752

Tungsten

powder, max. particle size 25 micron, weight 1000 g, purity 99.9%

Synonym(s):

W 006021, W

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9%

form

powder

manufacturer/tradename

Goodfellow 373-707-52

resistivity

4.9 μΩ-cm, 20°C

weight

1000 g

max. part. size

25 μm

bp

5660 °C (lit.)

mp

3410 °C (lit.)

density

19.3 g/mL at 25 °C (lit.)

SMILES string

[W]

InChI

1S/W

InChI key

WFKWXMTUELFFGS-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hi we are ordering GF37370752, Tungsten particle with max particle size of 25um. Is there a way that we could get the particle size distribution chart for my application development.

    1 answer
    1. This product is manufactered by Goodfellow under the product 373-707-52. This product is only offered in 25µm, there is no product distribution chart offered. Please see page 82 of this brochure from Goodfellow: https://www.goodfellow.co.kr/pub/media/pdf/Goodfellow-Product-List-Final.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service