Skip to Content
Merck
All Photos(2)

Key Documents

GF16286227

Silicon

disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%

Synonym(s):

Silicon, SI003051

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si
CAS Number:
Molecular Weight:
28.09
MDL number:
UNSPSC Code:
12141911
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

100%

form

foil

manufacturer/tradename

Goodfellow 162-862-27

diam. × thickness

15.9 mm × 0.38 mm

bp

2355 °C (lit.)

mp

1410 °C (lit.)

density

2.33 g/mL at 25 °C (lit.)

SMILES string

[Si]

InChI

1S/Si

InChI key

XUIMIQQOPSSXEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emil Rudobeck et al.
Radiation research, 181(4), 407-415 (2014-03-15)
An unavoidable complication of space travel is exposure to radiation consisting of high-energy charged particles (HZE), such as Fe and Si nuclei. HZE radiation can affect neuronal functions at the level of the synapse or neuronal soma without inducing significant
Dean G Johnson et al.
Advances in chronic kidney disease, 20(6), 508-515 (2013-11-12)
The development of wearable or implantable technologies that replace center-based hemodialysis (HD) hold promise to improve outcomes and quality of life for patients with ESRD. A prerequisite for these technologies is the development of highly efficient membranes that can achieve
Keith R Martin
Metal ions in life sciences, 13, 451-473 (2014-01-29)
Silicon is the second most abundant element in nature behind oxygen. As a metalloid, silicon has been used in many industrial applications including use as an additive in the food and beverage industry. As a result, humans come into contact
David J Savage et al.
Current opinion in pharmacology, 13(5), 834-841 (2013-07-13)
Biomedical applications of porous silicon include drug delivery, imaging, diagnostics and immunotherapy. This review summarizes new silicon particle fabrication techniques, dynamics of cellular transport, advances in the multistage vector approach to drug delivery, and the use of porous silicon as
Yanli Wang et al.
Advanced materials (Deerfield Beach, Fla.), 25(37), 5177-5195 (2013-07-06)
Semiconducting silicon nanowires (SiNWs) represent one of the most interesting research directions in nanoscience and nanotechnology, with capabilities of realizing structural and functional complexity through rational design and synthesis. The exquisite control of chemical composition, structure, morphology, doping, and assembly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service