Skip to Content
Merck
All Photos(2)

Key Documents

GF10746862

Bismuth

foil, not light tested, 150x150mm, thickness 0.02mm, permanent polyester support, 99.97%

Synonym(s):

Bismuth, BI000150

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi
CAS Number:
Molecular Weight:
208.98
MDL number:
UNSPSC Code:
12141706
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.97%

form

foil

manufacturer/tradename

Goodfellow 107-468-62

resistivity

129 μΩ-cm, 20°C

L × W × thickness

150 mm × 150 mm × 0.02 mm

bp

1560 °C (lit.)

mp

271 °C (lit.)

density

9.8 g/mL at 25 °C (lit.)

SMILES string

[Bi]

InChI

1S/Bi

InChI key

JCXGWMGPZLAOME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge A R Salvador et al.
Molecules (Basel, Switzerland), 16(4), 2884-2913 (2011-04-06)
Steroid and terpene chemistry still have a great impact on medicinal chemistry. Therefore, the development of new reactions or "greener" processes in this field is a contemporaneous issue. In this review, the use of bismuth(III) salts, as "ecofriendly" reagents/catalysts, on
Hong Lu et al.
European journal of gastroenterology & hepatology, 25(10), 1134-1140 (2013-06-20)
Antimicrobial resistance has continued to undermine many popular anti-Helicobacter pylori therapies. Antibiotic resistance to commonly used anti-H. pylori drugs in China has increased markedly, making China an ideal site to identify regimens that remain effective despite widespread antimicrobial resistance. Bismuth
Syntheses of biodegradable and biocompatible polymers by means of bismuth catalysts.
Hans R Kricheldorf
Chemical reviews, 109(11), 5579-5594 (2009-09-26)
Jorge A R Salvador et al.
Future medicinal chemistry, 4(11), 1495-1523 (2012-08-04)
In recent years, the chemical potential of bismuth and bismuth compounds has been actively exploited. Bismuth salts are known for their low toxicity, making them potential valuable reagents for large-scale synthesis, which becomes more obvious when dealing with products such
Bismuth in medicine.
Hongzhe Sun et al.
Metal ions in biological systems, 41, 333-378 (2004-06-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service