Key Documents
ESYNTH021
Sigma-Aldrich
SynLectro Nickel Foam Electrode
size 3 mm × 20 mm × 60 mm
Synonym(s):
Nickel Foam Electrochemical Electrode
About This Item
UNSPSC Code:
81141502
NACRES:
NB.61
Pricing and availability is not currently available.
size
3 mm × 20 mm × 60 mm
General description
SynLectro Nickel Foam Electrode is a 3x20x60 mm electrode designed for use with the SynLectro Electrolysis Platform for synthetic electrochemistry.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?
    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
      Helpful?
  2. How can I determine the shelf life / expiration / retest date of this product?
    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
      Helpful?
  3. Please, the density in g/cm3 is?
    1 answer
    1. The density is not determined for this product. Please note that this is not a batch-managed product designed to be used with the SynLectro™ Electrolysis Platform for synthetic electrochemistry. Unfortunately, the only information on the dimensions are the ones provided which are 3 mm × 20 mm × 60 mm.
      Helpful?