E33203
Ethyl isobutyrylacetate
95%
Synonym(s):
Ethyl 4-methyl-3-oxopentanoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCOCH2COOC2H5
CAS Number:
Molecular Weight:
158.19
Beilstein:
636726
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.425 (lit.)
bp
173 °C (lit.)
mp
−9 °C (lit.)
density
0.98 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)CC(=O)C(C)C
InChI
1S/C8H14O3/c1-4-11-8(10)5-7(9)6(2)3/h6H,4-5H2,1-3H3
InChI key
XCLDSQRVMMXWMS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
111.2 °F - closed cup
Flash Point(C)
44 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service