Skip to Content
Merck
All Photos(3)

Documents

E15701

Sigma-Aldrich

Ethyl butyrate

99%

Synonym(s):

Ethyl butanoate, Butyric acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(O)OC2H5
CAS Number:
Molecular Weight:
116.16
Beilstein:
506331
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

vapor pressure

15.5 mmHg ( 25 °C)

Assay

99%

form

liquid

autoignition temp.

865 °F

refractive index

n20/D 1.392 (lit.)

bp

120 °C (lit.)

mp

−93 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CCCC(=O)OCC

InChI

1S/C6H12O2/c1-3-5-6(7)8-4-2/h3-5H2,1-2H3

InChI key

OBNCKNCVKJNDBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl butyrate is a short-chain, fruit-flavored ester mainly generally used in food and pharmaceutical products.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esterification activity and operational stability of Candida rugosa lipase immobilized in polyurethane foams in the production of ethyl butyrate.
Pires-Cabral P, et al.
Biochemical Engineering Journal, 48(2), 246-252 (2010)
Short-chain flavour ester synthesis by immobilized lipase in organic media.
Manjon A, et al.
Biotechnology Letters, 13(5), 339-344 (1991)
Suhyoung Bahk et al.
BMC biology, 14(1), 83-83 (2016-10-01)
Like most animals, insects rely on their olfactory systems for finding food and mates and in avoiding noxious chemicals and predators. Most insect olfactory neurons express an odorant-specific odorant receptor (OR) along with Orco, the olfactory co-receptor. Orco binds ORs
Dieter Wicher et al.
Nature, 452(7190), 1007-1011 (2008-04-15)
From worm to man, many odorant signals are perceived by the binding of volatile ligands to odorant receptors that belong to the G-protein-coupled receptor (GPCR) family. They couple to heterotrimeric G-proteins, most of which induce cAMP production. This second messenger
D Labbe et al.
Chemical senses, 32(3), 205-214 (2006-11-03)
The impact of olfactory perception on sweetness was explored in a model solution using odorants at subthreshold concentrations. First, the impact of 6 odorants, previously described in the literature as congruent with sweetness, was investigated at suprathreshold level in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service