DWK212162806
Duran® Erlenmeyer narrow-neck flasks
flask capacity (125 mL)
Synonym(s):
212162806
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.17
Recommended Products
material
borosilicate glass 3.3 flask
flat
Sterilization Compatibility
autoclavable compatible
feature
without cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 212162806
flask capacity
(125 mL)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Duran® Erlenmeyer narrow-neck conical flasks are ideal for mixing.
- Made from borosilicate glass 3.3 for better clarity, chemical resistance, and minimal thermal expansion.
- Heavy and uniform wall thickness facilitates its usage under extreme temperature fluctuations.
- Easy-to-read scale with graduation marks in highly durable white enamel.
- Marking spot for sample identification.
Easy mixing due to conical form. Fired-on, highly durable white ceramic print.
Other Notes
Browse all Erlenmeyer flasks
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service