Skip to Content
Merck
All Photos(1)

Key Documents

D76722

Sigma-Aldrich

1,4-Dicyanobenzene

98%

Synonym(s):

Terephthalonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CN)2
CAS Number:
Molecular Weight:
128.13
Beilstein:
1072210
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

221-225 °C (lit.)

SMILES string

N#Cc1ccc(cc1)C#N

InChI

1S/C8H4N2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H

InChI key

BHXFKXOIODIUJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christian Ochsenfeld et al.
Solid state nuclear magnetic resonance, 22(2-3), 128-153 (2002-12-10)
A study of a host-guest system consisting of a naphthalene-spaced tweezer with a 1,4 dicyanobenzene guest molecule is presented. The complex is investigated using a combination of quantum-chemical calculations and solid-state NMR experiments. The advantages of such an approach are
Suchandra Chatterjee et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(1-2), 97-102 (2003-12-13)
With the advent of spin chemistry, magnetic field effect (MFE) on exciplex luminescence has emerged as an important domain of research. MFE is a diffusion controlled phenomenon and hence is solvent dielectric (epsilon) dependent. It maximizes at a particular epsilon
T Sengupta et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57A(5), 1103-1110 (2001-05-26)
The exciplex between all-s-trans-1,4-diphenylbuta-1,3-diene and 1,4-dicyanobenzene has been studied by steady state fluorescence along with the magnetic field effect (MFE) and compared with the other alpha,omega-diphenyl polyenes. The exciplex formation and magnetic field effect are dictated by the chain length
Felix Eberle et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(13), 2951-2956 (2010-08-18)
Self-assembled monolayers of 1,4-dicyanobenzene on Au(111) electrodes are studied by cyclic voltammetry, in-situ STM and ex-situ XPS. High-resolution STM images reveal a long-range order of propeller-like assemblies each of which consists of three molecules, all lying flat on the gold
J Pukkila et al.
The Analyst, 114(9), 1009-1012 (1989-09-01)
A method for the determination of emissions in workroom air as a result of the thermal degradation of polyester coating powders is described. The method is based on the determination of certain marker compounds only. Polyester coating powders were degraded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service