All Photos(2)
About This Item
Linear Formula:
Cl2C6H3CH2NH2
CAS Number:
Molecular Weight:
176.04
Beilstein:
775667
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.578 (lit.)
bp
83-84 °C/5 mmHg (lit.)
density
1.308 g/mL at 25 °C (lit.)
SMILES string
NCc1ccc(Cl)cc1Cl
InChI
1S/C7H7Cl2N/c8-6-2-1-5(4-10)7(9)3-6/h1-3H,4,10H2
InChI key
SJUKJZSTBBSGHF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
William C Jolin et al.
Environmental science & technology, 50(15), 8196-8204 (2016-07-06)
Column chromatography was evaluated as a method to obtain organic cation sorption isotherms for environmental solids while using the peak skewness to identify the linear range of the sorption isotherm. Custom packed HPLC columns and standard batch sorption techniques were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service