D23408
2,4-Diamino-6-phenyl-1,3,5-triazine
97%
Synonym(s):
6-Phenyl-1,3,5-triazine-2,4-diamine, Benzoguanamine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C9H9N5
CAS Number:
Molecular Weight:
187.20
Beilstein:
153223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
powder
mp
219-221 °C (lit.)
SMILES string
Nc1nc(N)nc(n1)-c2ccccc2
InChI
1S/C9H9N5/c10-8-12-7(13-9(11)14-8)6-4-2-1-3-5-6/h1-5H,(H4,10,11,12,13,14)
InChI key
GZVHEAJQGPRDLQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Eagambaram Murugan et al.
ACS omega, 3(10), 13685-13693 (2018-11-10)
Four different novel magnetic core-shell nanocomposites stabilized with Au/Pd nanoparticles (NPs) were prepared by a simple procedure and demonstrated their catalytic activity for effective cleavage of pBR322 DNA. Initially, the Fe3O4-poly(styrene-divinylbenzene-vinylbenzyl chloride) (ST-DVB-VBC) matrix functionalized with 3-aminobenzoic acid was prepared
Bochuan Liu et al.
Chemosphere, 189, 115-122 (2017-09-22)
The enhancement of g-C
K Sivakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 200, 212-225 (2018-04-25)
The inclusion complex of β-cyclodextrin (β-CD) with benzoguanamine (BGA) has been investigated in three states. UV-Visible and fluorescence spectral techniques are used in liquid state. FTIR, NMR and MASS techniques are used in solid state and virtual state studies are
Assessment of the Impact of Accelerated Migration Testing for Coated Food Cans Using Food Simulants.
Rafael Paseiro-Cerrato et al.
Molecules (Basel, Switzerland), 24(17) (2019-08-31)
In this study, an accelerated migration test on food can coatings into food simulants was investigated. Food simulants covering a wide range of polarity were used to conduct migration tests at 60 °C with storage times ranging from 4 h
Qian Cao et al.
Polymers, 12(8) (2020-07-28)
High potential of emission chemistry has been visualized in many fields, from sensors and imaging to displays. In general, conjugated polymers are the top rankers for such chemistry, despite the fact that they bring solubility problems, high expenses, toxicity and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service