Skip to Content
Merck
All Photos(1)

Documents

D206555

Sigma-Aldrich

Diphenyl phosphoryl chloride

99%

Synonym(s):

Diphenyl chlorophosphate, Diphenyl phosphorochloridate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)2POCl
CAS Number:
Molecular Weight:
268.63
Beilstein:
654130
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10 mmHg ( 190 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.55 (lit.)

bp

314-316 °C/272 mmHg (lit.)

density

1.296 g/mL at 25 °C (lit.)

SMILES string

ClP(=O)(Oc1ccccc1)Oc2ccccc2

InChI

1S/C12H10ClO3P/c13-17(14,15-11-7-3-1-4-8-11)16-12-9-5-2-6-10-12/h1-10H

InChI key

BHIIGRBMZRSDRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphorylation reagent for protected serine and pyranoses.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbohydrate Research, 223, 169-169 (1992)
Tetrahedron Letters, 34, 2461-2461 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service