Skip to Content
Merck
All Photos(1)

Key Documents

D139459

Sigma-Aldrich

4-(Dimethylamino)benzoic acid

98%, for peptide synthesis

Synonym(s):

N,N-Dimethyl-4-aminobenzoic acid, N,N-Dimethyl-p-aminobenzoic acid, p-(Dimethylamino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CO2H
CAS Number:
Molecular Weight:
165.19
Beilstein:
386867
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

product name

4-(Dimethylamino)benzoic acid, 98%

Quality Level

Assay

98%

form

powder and chunks

reaction suitability

reaction type: solution phase peptide synthesis

mp

241-243 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CN(C)c1ccc(cc1)C(O)=O

InChI

1S/C9H11NO2/c1-10(2)8-5-3-7(4-6-8)9(11)12/h3-6H,1-2H3,(H,11,12)

InChI key

YDIYEOMDOWUDTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Hino et al.
Journal of analytical toxicology, 24(3), 165-169 (2000-04-25)
A sensitive and reliable method was developed for the determination of tetracaine and its metabolite, p-butylaminobenzoic acid, in human samples. Tetracaine and the metabolite, effectively extracted using a liquid-liquid extraction procedure from 0.5 g of sample, were analyzed by gas
Y B Jiang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56(4), 623-627 (2000-05-04)
Photoinduced intramolecular charge transfer (ICT) of p-N,N-dimethylaminobenzoic acid (DMABOA) in AOT/cyclohexane/H2O reverse micelle was investigated and compared with that in CTAB/1-heptanol/H2O reverse micelle. It is proposed that the DMABOA molecule exists at the AOT reverse micelle water pool interface with
Xiaoming Xu et al.
Dental materials : official publication of the Academy of Dental Materials, 22(11), 1014-1023 (2005-12-28)
The aims of this study were to formulate a novel fluoride-releasing dental composite and to evaluate its mechanical properties, fluoride release and recharge capabilities, water sorption and solubility. A fluoride-releasing dimethacrylate monomer containing a ternary zirconium fluoride chelate was synthesized.
P V Argade et al.
Biochemistry, 23(2), 299-304 (1984-01-17)
The imidazole of chromophoric p-(dimethylamino)benzoic acid, DABIm, reacts with the serine protease alpha-chymotrypsin in the pH range of 4-7 to form a stable acyl intermediate that gives very good resonance-enhanced Raman spectra. The resonance Raman and absorption spectra of the
Xinglin Guo et al.
Dental materials : official publication of the Academy of Dental Materials, 24(6), 824-831 (2007-11-30)
The purpose of this study was to evaluate the effects of photoinitiator type and water content on the polymerization rate (Rp) and degree of conversion (DC) of a model BisGMA/HEMA-based resin. The comonomer mixture consisted of BisGMA/HEMA (60/40 by weight).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service