D103705
Diglycolic anhydride
technical grade, 90%
Synonym(s):
1,4-Dioxane-2,6-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H4O4
CAS Number:
Molecular Weight:
116.07
Beilstein:
112527
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
grade
technical grade
Quality Level
Assay
90%
form
crystals
bp
240-241 °C (lit.)
mp
92-93 °C (lit.)
SMILES string
O=C1COCC(=O)O1
InChI
1S/C4H4O4/c5-3-1-7-2-4(6)8-3/h1-2H2
InChI key
PIYNUZCGMLCXKJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jing-Min Liu et al.
Journal of hazardous materials, 344, 220-229 (2017-10-19)
Herein, for the first time, the typical porous Covalent Organic Frameworks (COFs) CTpBD with superior chemical stability and large surface area were applied as sorbents for solid phase extraction of trace ions via flow injection followed by inductively coupled plasma
Jiao Feng et al.
Science advances, 5(2), eaau5148-eaau5148 (2019-02-23)
The clinical use of endogenous neuropeptides has historically been limited due to pharmacokinetic issues, including plasma stability and blood-brain barrier permeability. In this study, we show that the rapidly metabolized Leu-enkephalin (LENK) neuropeptide may become pharmacologically efficient owing to a
Ying Gu et al.
Biosensors & bioelectronics, 128, 129-136 (2019-01-19)
For the determination of dopamine (DA) in serum samples, a quenching-type electrochemiluminescence sensor (MIECLS) was constructed in this study. Upconversion nanoparticles (UCNPs) enhanced by covalent organic frameworks (COFs)-based hybrid and oligoaniline-crosslinked gold nanoparticles (AuNPs) imprinting recognition sites were introduced in
Daniele Vinciguerra et al.
Journal of controlled release : official journal of the Controlled Release Society, 295, 223-236 (2019-01-07)
Well-defined, heterotelechelic polymer prodrugs for combination therapy were synthesized by using a combination of the "drug-initiated" nitroxide-mediated polymerization from a gemcitabine-alkoxyamine initiator and the nitroxide exchange reaction using TEMPO-bearing drugs to end-cap the drug-polymer chain-end by a second drug. This
Baolong Xie et al.
Journal of inorganic biochemistry, 171, 67-75 (2017-04-04)
Aggregation of amyloid β-proteins (Aβ) induced by Cu
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service