Key Documents
CLS5600100
Pyrex® Lifetime-Red volumetric flask with snap cap
capacity 100 mL
Synonym(s):
clear flasks, pyrex flask, volumetric flasks, Corning® 5600
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
long
sterility
non-sterile
feature
closure type Snap cap
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5600-100
tol. measuring range
0.08 mL
capacity
100 mL
General description
Capacity mark is fine line-etched through narrow, red band into clear glass.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)